Iodixanol Related Compound D
Synonym(s):
Iopentol, 5-[Acetyl(2-hydroxy-3-methylpropyl)amino]-N,N′-bis(2,3-dihydroxypropyl)-2,4,6-triiodo-1,3-benzenedicarboxamide
Empirical Formula (Hill Notation):
C20H28I3N3O9
CAS Number:
Molecular Weight:
835.16
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing