Indium(III) oxide
Synonym(s):
Diindium trioxide, Indium sesquioxide, Indium(III) oxide
Empirical Formula (Hill Notation):
In2O3
CAS Number:
Molecular Weight:
277.63
Products
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Application
Reagent Type
Core