![Imiquimod Related Compound C Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/images/124/976/69a991b7-2f59-4924-84d6-b4a754c7d3f4/640/69a991b7-2f59-4924-84d6-b4a754c7d3f4.jpg)
Imiquimod Related Compound C
![Imiquimod Related Compound C Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/images/124/976/69a991b7-2f59-4924-84d6-b4a754c7d3f4/640/69a991b7-2f59-4924-84d6-b4a754c7d3f4.jpg)
Synonym(s):
4-Chloro-1-isobutyl-1H-imidazo[4,5-c]quinoline, NSC 672965
Empirical Formula (Hill Notation):
C14H14ClN3
CAS Number:
Molecular Weight:
259.73
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2