![Imiquimod Pharmaceutical Secondary Standard; Certified Reference Material, certified reference material, pharmaceutical secondary standard, pkg of 500 mg](/deepweb/assets/sigmaaldrich/product/structures/239/355/39eea20d-5d3b-4ac6-b475-3aa80b26ee9b/640/39eea20d-5d3b-4ac6-b475-3aa80b26ee9b.png)
Imiquimod
![Imiquimod Pharmaceutical Secondary Standard; Certified Reference Material, certified reference material, pharmaceutical secondary standard, pkg of 500 mg](/deepweb/assets/sigmaaldrich/product/structures/239/355/39eea20d-5d3b-4ac6-b475-3aa80b26ee9b/640/39eea20d-5d3b-4ac6-b475-3aa80b26ee9b.png)
Synonym(s):
1-(2-Methylpropyl)-1H-imidazole[4,5-c]quinoline-4-amine, Imiquimod
Empirical Formula (Hill Notation):
C14H16N4
CAS Number:
Molecular Weight:
240.30
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Agency
Format
Research Area
Quality Segment
Application
Package Size
API Family
Special Grade
Showing 1-4 of 4
Compare
Product Number
Description
Pricing