Skip to Content
Merck
Hyperforin (dicyclohexylammonium) salt

Hyperforin (dicyclohexylammonium) salt

Hyperforin (dicyclohexylammonium) salt
Synonym(s):
Hyp-DCHA, Hyperforin-DCHA
Empirical Formula (Hill Notation):
C35H52O4·C12H23N
CAS Number:
Molecular Weight:
718.10

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing