Skip to Content
Merck
Harpagoside primary reference standard

Harpagoside

Harpagoside primary reference standard
Synonym(s):
(1S ,4aS ,5R ,7S ,7aS )-1,4a,5,6,7,7a-Hexahydro-4a,5-dihydroxy-7-methyl-7-[[(2E )-1-oxo-3-phenyl-2-propen-1-yl]oxy]cyclopenta[c ]pyran-1-yl β-D-glucopyranoside
Empirical Formula (Hill Notation):
C24H30O11
CAS Number:
Molecular Weight:
494.49
EC Number:

Products

Brand
Color
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique