Skip to Content
Merck
Granzyme B human recombinant, expressed in insect cells, ≥90% (SDS-PAGE)

Granzyme B human

Granzyme B human recombinant, expressed in insect cells, ≥90% (SDS-PAGE)
Synonym(s):
Granzyme B human, Cytotoxic T-lymphocyte proteinase 2, Granzyme-2, GranzymeB, Lymphocyte protease, SECT, T-cell serine protease 1-3E

Products

Product Category
Brand
Biological Source
Gene Alias
Physical Form
Purity
Recombinant Host
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing