Gramine
Synonym(s):
3-(Dimethylaminomethyl)indole, Donaxine, NSC 16892
Empirical Formula (Hill Notation):
C11H14N2
CAS Number:
Molecular Weight:
174.24
Products
Product Category
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Product Line
Purity
Format
Markush Class
Markush Group
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing