![Gossypin phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/200/592/f789a80c-6f0c-40ec-830f-7a1f28d39158/640/f789a80c-6f0c-40ec-830f-7a1f28d39158.png)
Gossypin
![Gossypin phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/200/592/f789a80c-6f0c-40ec-830f-7a1f28d39158/640/f789a80c-6f0c-40ec-830f-7a1f28d39158.png)
Synonym(s):
2-(3,4-Dihydroxyphenyl)-8-(β-D-glucopyranosyloxy)-3,5,7-trihydroxy- 4H-1-benzopyran-4-one, 3,3′,4′,5,7,8-Hexahydroxyflavone 8-glucoside, Gossypetin 8-glucoside
Empirical Formula (Hill Notation):
C21H20O13
CAS Number:
Molecular Weight:
480.38
Products
Formula Weight
Manufacturer
Product Line
Purity
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing