Skip to Content
Merck
Gly-Gln monohydrate ≥97% (HPLC)

Gly-Gln monohydrate

Gly-Gln monohydrate ≥97% (HPLC)
Synonym(s):
Glycyl-L-glutamine
Empirical Formula (Hill Notation):
C7H13N3O4 · H2O
CAS Number:
Molecular Weight:
221.21

Products

Product Category
Brand
Formula Weight
Purity
Research Area
Showing 1-1 of 1
Compare
Product Number
Description
Pricing