Skip to Content
Merck
Glycyrrhizic acid ammonium salt analytical standard, suitable for HPLC

Glycyrrhizic acid ammonium salt

Glycyrrhizic acid ammonium salt analytical standard, suitable for HPLC
Empirical Formula (Hill Notation):
C42H62O16 · NH3
CAS Number:
Molecular Weight:
839.96
EC Number:

Products

Product Category
Brand
Formula Weight
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing