![Glyburide Related Compound A United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/101/378/bad0e801-dad2-49ac-9930-0d63d876c939/640/bad0e801-dad2-49ac-9930-0d63d876c939.png)
Glyburide Related Compound A
![Glyburide Related Compound A United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/101/378/bad0e801-dad2-49ac-9930-0d63d876c939/640/bad0e801-dad2-49ac-9930-0d63d876c939.png)
Synonym(s):
5-Chloro-2-methoxy-N-[2-(4-sulfamoylphenyl)ethyl]benzamide, 4-[2-(5-Chloro-2-methoxybenzamido)ethyl]phenylsulfonamide
Linear Formula:
ClC6H3(OCH3)CONHCH2C6H4SO2NH2
CAS Number:
Molecular Weight:
368.84
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing