
Fmoc N-hydroxysuccinimide ester

Synonym(s):
9-Fluorenylmethyl N-succinimidyl carbonate, N-(9-Fluorenylmethoxycarbonyloxy)succinimide, Fmoc-OSu
Empirical Formula (Hill Notation):
C19H15NO5
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing