
Flurandrenolide

Synonym(s):
6α-Fluoro-11β,16α,17,21-tetrahydroxypregn-4-ene-3,20-dione 16,17-acetonide, Fludroxycortide
Empirical Formula (Hill Notation):
C24H33FO6
CAS Number:
Molecular Weight:
436.51
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing