![Fluorouracil Related Compound A certified reference material, pharmaceutical secondary standard](/deepweb/assets/sigmaaldrich/product/structures/415/889/6e64b93a-548a-4331-8c38-cdf05e3b3fab/640/6e64b93a-548a-4331-8c38-cdf05e3b3fab.png)
Fluorouracil Related Compound A
![Fluorouracil Related Compound A certified reference material, pharmaceutical secondary standard](/deepweb/assets/sigmaaldrich/product/structures/415/889/6e64b93a-548a-4331-8c38-cdf05e3b3fab/640/6e64b93a-548a-4331-8c38-cdf05e3b3fab.png)
Synonym(s):
Barbituric acid, 2,4,6-Trihydroxypyrimidine, Malonylurea
Empirical Formula (Hill Notation):
C4H4N2O3
CAS Number:
Molecular Weight:
128.09
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2