Fluorescein Phosphoramidite
Synonym(s):
Fluorescein amidite
Empirical Formula (Hill Notation):
C46H58N3O10P
Molecular Weight:
843.94
Products
Product Category
Brand
Formula Weight
Product Line
Showing 1-2 of 2
Compare
Product Number
Description
Pricing