Skip to Content
Merck
Flumetsulam PESTANAL®, analytical standard

Flumetsulam

Flumetsulam PESTANAL®, analytical standard
Synonym(s):
2′,6′-Difluoro-5-methyl-[1,2,4]triazolo[1,5-a]pyrimidine-2-sulfonanilide, N-(2,6-Difluorophenyl)-5-methyl-[1,2,4]triazolo[1,5-a]pyrimidine-2-sulfonamide
Empirical Formula (Hill Notation):
C12H9F2N5O2S
CAS Number:
Molecular Weight:
325.29

Products

Product Category
Brand
Formula Weight
Product Line
Purity
Format
Chemical Composition
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing