Fluconazole Related Compound B
Synonym(s):
2-(4-Fluorophenyl)-1,3-bis(1H-1,2,4-triazol-1-yl)propan-2-ol
Empirical Formula (Hill Notation):
C13H13FN6O
CAS Number:
Molecular Weight:
288.28
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2