Skip to Content
Merck
Fenofibrate Related Compound B certified reference material, pharmaceutical secondary standard

Fenofibrate Related Compound B

Fenofibrate Related Compound B certified reference material, pharmaceutical secondary standard
Synonym(s):
2-[4-(4-Chlorobenzoyl)phenoxy]-2-methylpropionic acid, Fenofibric acid
Empirical Formula (Hill Notation):
C17H15ClO4
CAS Number:
Molecular Weight:
318.75

Products

Brand
Formula Weight
Gene Alias
Manufacturer
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2