Skip to Content
Merck
Ethyl O-(2-mesitylenesulfonyl)acethydroxamate purum, ≥97.0% (HPLC)

Ethyl O-(2-mesitylenesulfonyl)acethydroxamate

Ethyl O-(2-mesitylenesulfonyl)acethydroxamate purum, ≥97.0% (HPLC)
Synonym(s):
O-(2-Mesitylenesulfonyl)acethydroxamic acid ethyl ester
Linear Formula:
2,4,6-(CH3)3C6H2SO2ON=C(CH3)OC2H5
CAS Number:
Molecular Weight:
285.36
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing