Skip to Content
Merck
Ethyl 2-(bromomethyl)acrylate 98%

Ethyl 2-(bromomethyl)acrylate

Ethyl 2-(bromomethyl)acrylate 98%
Linear Formula:
CH2=C(CH2Br)CO2CH2CH3
CAS Number:
Molecular Weight:
193.04

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Purity
Markush Class
Markush Group
Monomer Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing