Skip to Content
Merck
Etamsylate ≥95% (HPLC)

Etamsylate

Etamsylate ≥95% (HPLC)
Synonym(s):
Etamsylate, 2,5-Dihydroxybenzenesulfonic acid diethylamine salt, CID 17506, Diethylammonium 2,5-dihydroxybenzenesulfonate
Empirical Formula (Hill Notation):
C10H17NO5S
CAS Number:
Molecular Weight:
263.31

Products

Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Format
Research Area
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing