![EPA 8260 Internal Standards Mix certified reference material, TraceCERT®, 2000 μg/mL each component in methanol](/deepweb/assets/sigmaaldrich/product/images/106/253/1101c775-293a-4ee1-982f-5e8de0407618/640/1101c775-293a-4ee1-982f-5e8de0407618.jpg)
EPA 8260 Internal Standards Mix
![EPA 8260 Internal Standards Mix certified reference material, TraceCERT®, 2000 μg/mL each component in methanol](/deepweb/assets/sigmaaldrich/product/images/106/253/1101c775-293a-4ee1-982f-5e8de0407618/640/1101c775-293a-4ee1-982f-5e8de0407618.jpg)
Synonym(s):
EPA 8260A Internal Standards Mix
Products
Brand
Product Line
Agency
Format
Quality Segment
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing