Skip to Content
Merck
ε-Caprolactone 97%

ε-Caprolactone

ε-Caprolactone 97%
Synonym(s):
2-Oxepanone, 6-Caprolactone monomer, 6-Hexanolactone, ε-Caprolactone
Empirical Formula (Hill Notation):
C6H10O2
CAS Number:
Molecular Weight:
114.14

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Monomer Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing