dUTP
Synonym(s):
2′-Deoxyuridine 5′-triphosphate sodium salt solution, dUTP sodium salt
Empirical Formula (Hill Notation):
C9H15N2O14P3 · xNa+
CAS Number:
Molecular Weight:
468.14 (free acid basis)
Products
Formula Weight
Manufacturer
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing