![Docetaxel impurity E European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/108/818/6a348c2e-53f9-444a-9b44-5124e3398fbf/640/6a348c2e-53f9-444a-9b44-5124e3398fbf.png)
Docetaxel impurity E
![Docetaxel impurity E European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/108/818/6a348c2e-53f9-444a-9b44-5124e3398fbf/640/6a348c2e-53f9-444a-9b44-5124e3398fbf.png)
Synonym(s):
10-Deacetylbaccatin III
Empirical Formula (Hill Notation):
C29H36O10
CAS Number:
Molecular Weight:
544.59
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing