DMT-2′O-TC-rA(ib) Phosphoramidite
Synonym(s):
DMT-2′O-TC-rA(ib) Amidite
Empirical Formula (Hill Notation):
C49H61N8O10PS2
CAS Number:
Molecular Weight:
1017.16
Products
Product Category
Brand
Formula Weight
Quality Segment
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing