Skip to Content
Merck
DL-Homophenylalanine 98%

DL-Homophenylalanine

DL-Homophenylalanine 98%
Synonym(s):
(±)-2-Amino-4-phenylbutyric acid
Linear Formula:
C6H5(CH2CH2)CH(NH2)CO2H
CAS Number:
Molecular Weight:
179.22

Products

Brand
Formula Weight
Melting Point (°C)
Purity
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Amino Acid Type
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing