(±)-Cotinine-(methyl-d3)
Synonym(s):
1-(Methyl-d3)-5-(3-pyridinyl)-2-pyrrolidinone, Deuterated (±)-cotinine solution
Empirical Formula (Hill Notation):
C10D3H9N2O
CAS Number:
Molecular Weight:
179.23
Products
Product Category
Brand
Boiling Point (°C)
Color
Formula Weight
Melting Point (°C)
Physical Form
Isotope
Markush Class
Markush Group
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing