Skip to Content
Merck
(±)-2-Octanol analytical standard

(±)-2-Octanol

(±)-2-Octanol analytical standard
Synonym(s):
Hexyl methyl carbinol
Linear Formula:
CH3(CH2)5CH(OH)CH3
CAS Number:
Molecular Weight:
130.23
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing