![Dipotassium clorazepate monohydrate European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/227/605/002e4d0e-380a-4fbd-ad69-5d456951de18/640/002e4d0e-380a-4fbd-ad69-5d456951de18.png)
Dipotassium clorazepate monohydrate
![Dipotassium clorazepate monohydrate European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/227/605/002e4d0e-380a-4fbd-ad69-5d456951de18/640/002e4d0e-380a-4fbd-ad69-5d456951de18.png)
Synonym(s):
Clorazepate dipotassium salt, 7-Chloro-2,3-dihydro-2,2-dihydroxy-5-phenyl-1H-1,4-benzodiazepine-3-carboxylic acid
Linear Formula:
C16H11ClN2O4K2
CAS Number:
Molecular Weight:
408.92
Products
Formula Weight
Manufacturer
API Family
Reference Material Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing