
Dimethylaminomethyl-polystyrene

Synonym(s):
Benzyldimethylamine polymer-bound, Copolymer of styrene and divinylbenzene, dimethylaminomethylated, Polystyrene crosslinked with divinylbenzene, dimethylaminomethylated
Linear Formula:
CH2N(CH3)2 - [CH2CH(C6H55)]n
Products
Brand
Chain Type
Reaction Type
Reactivity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing