Skip to Content
Merck
Diethylamine NONOate sodium salt hydrate ≥97% (NMR)

Diethylamine NONOate sodium salt hydrate

Diethylamine NONOate sodium salt hydrate ≥97% (NMR)
Synonym(s):
1,1-Diethyl-2-hydroxy-2-nitroso-hydrazine sodium, 2-(N,N-Diethylamino)-diazenolate 2-oxide sodium salt hydrate, DEA NONOate sodium salt hydrate, Diethylamine/nitric oxide complex sodium salt hydrate
Linear Formula:
(CH3CH2)2N–N(N=O)O- Na+ xH2O
CAS Number:
Molecular Weight:
155.13 (anhydrous basis)

Products

Product Category
Brand
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Showing 1-1 of 1