![Didymin phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/350/276/bcedb796-61e5-4598-b1d9-e06ac9b5a20d/640/bcedb796-61e5-4598-b1d9-e06ac9b5a20d.png)
Didymin
![Didymin phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/350/276/bcedb796-61e5-4598-b1d9-e06ac9b5a20d/640/bcedb796-61e5-4598-b1d9-e06ac9b5a20d.png)
Synonym(s):
(S)-5,7-Dihydroxy-4′-methoxyflavanone-7-β-rutinoside, Isosakuranetin 7-O-rutinoside, Neoponcirin
Empirical Formula (Hill Notation):
C28H34O14
CAS Number:
Molecular Weight:
594.56
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing