Skip to Content
Merck
D-Glucuronic acid sodium salt monohydrate 97.5-102.5% (non-aqueous titration)

D-Glucuronic acid sodium salt monohydrate

D-Glucuronic acid sodium salt monohydrate 97.5-102.5% (non-aqueous titration)
Synonym(s):
Sodium D-glucuronate
Empirical Formula (Hill Notation):
C6H9NaO7 · H2O
CAS Number:
Molecular Weight:
234.14
EC Number:

Products

Product Category
Brand
Biological Source
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing