D-Glucuronic acid
Synonym(s):
Glucosiduronic acid, D-Glucuronic acid
Empirical Formula (Hill Notation):
C6H10O7
CAS Number:
Molecular Weight:
194.14
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3