Dexmedetomidine hydrochloride
Synonym(s):
(+)-Medetomidine hydrochloride, (S)-Medetomidine hydrochloride, 4-[(S)-α,2,3-Trimethylbenzyl]imidazole monohydrochloride, 5-[(1S)-1-(2,3-Dimethylphenyl)ethyl]-1H-imidazole hydrochloride, Dexmedetomidine hydrochloride
Empirical Formula (Hill Notation):
C13H16N2 · HCl
CAS Number:
Molecular Weight:
236.74
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Purity
Agency
Format
Research Area
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade