Skip to Content
Merck
Deoxycholic Acid (Desoxycholic Acid) United States Pharmacopeia (USP) Reference Standard

Deoxycholic Acid (Desoxycholic Acid)

Deoxycholic Acid (Desoxycholic Acid) United States Pharmacopeia (USP) Reference Standard
Synonym(s):
Deoxycholic acid, 3α,12α-Dihydroxy-5β-cholanic acid, 7-Deoxycholic acid, Desoxycholic acid
Empirical Formula (Hill Notation):
C24H40O4
CAS Number:
Molecular Weight:
392.57

Products

Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1