Debrisoquine sulfate
Synonym(s):
3,4-Dihydro-2(1H)-isoquinolinecarboximidamide, Debrisoquin sulfate, Ro 5-33071
Linear Formula:
C20H26N6 · H2SO4
CAS Number:
Molecular Weight:
448.54
Products
Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing