
Darutoside

Synonym(s):
(2R,4aS,4bR,7S,10aS)-7-[(1R)-1,2-Dihydroxyethyl]-1,2,3,4,4a,4b,5,6,7,9,10,10a-dodecahydro-1,1,4a,7-tetramethyl-2-phenanthrenyl-β-D-glucopyranoside, Darutigenol 3-β-D-glucoside
Empirical Formula (Hill Notation):
C26H44O8
CAS Number:
Molecular Weight:
484.62
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing