Skip to Content
Merck
D-Arabino-1,4-lactone ≥97.0% (HPLC)

D-Arabino-1,4-lactone

D-Arabino-1,4-lactone ≥97.0% (HPLC)
Synonym(s):
D-Arabinonic acid γ-lactone, D-Arabinono-1,4-lactone, D-Arabonic acid-1,4-lactone
Empirical Formula (Hill Notation):
C5H8O5
CAS Number:
Molecular Weight:
148.11

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing