
D-Amino Acid Oxidase Inhibitor III

Synonym(s):
3-Methylpyrazole-5-carboxylic acid, 5-Methylpyrazole-3-carboxylic acid, AS057278, MPC
Empirical Formula (Hill Notation):
C5H6N2O2
CAS Number:
Molecular Weight:
126.11
Products
Brand
Formula Weight
Melting Point (°C)
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing