Cyclopentolate hydrochloride
Synonym(s):
2-Dimethylaminoethyl 1-hydroxy-α-phenylcyclopentaneacetate
Empirical Formula (Hill Notation):
C17H25NO3 · HCl
CAS Number:
Molecular Weight:
327.85
Products
Formula Weight
Gene Alias
Manufacturer
Physical Form
Format
Application
API Family
Special Grade
Showing 1-3 of 3