Skip to Content
Merck
Cyanocobalamin (crystallline) United States Pharmacopeia (USP) Reference Standard

Cyanocobalamin (crystallline)

Cyanocobalamin (crystallline) United States Pharmacopeia (USP) Reference Standard
Synonym(s):
Vitamin B12, α-(5,6-Dimethylbenzimidazolyl)cyanocobamide, CN-Cbl, Cyanocob(III)alamin, Cyanocobalamin
Empirical Formula (Hill Notation):
C63H88CoN14O14P
CAS Number:
Molecular Weight:
1355.37

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1