![Ciprofloxacin HCl Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/structures/374/908/7a74a36e-fb89-4748-9c8d-92508e177129/640/7a74a36e-fb89-4748-9c8d-92508e177129.png)
Ciprofloxacin HCl
![Ciprofloxacin HCl Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/structures/374/908/7a74a36e-fb89-4748-9c8d-92508e177129/640/7a74a36e-fb89-4748-9c8d-92508e177129.png)
Synonym(s):
Ciprofloxacin hydrochloride monohydrate
Empirical Formula (Hill Notation):
C17H18FN3O3 · HCl · H2O
CAS Number:
Molecular Weight:
385.82
Products
Brand
Formula Weight
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing