![Cinmethylin PESTANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/362/628/ef4fa114-d3e2-480e-9937-7010f6c03475/640/ef4fa114-d3e2-480e-9937-7010f6c03475.png)
Cinmethylin
![Cinmethylin PESTANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/362/628/ef4fa114-d3e2-480e-9937-7010f6c03475/640/ef4fa114-d3e2-480e-9937-7010f6c03475.png)
Synonym(s):
exo-1-Methyl-4-(1-methylethyl)-2-[(2- methylphenyl)methoxy]-7-oxabicyclo[2.2.1]heptane
Empirical Formula (Hill Notation):
C18H26O2
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2