Skip to Content
Merck
Cholesteryl phenylacetate ≥98% (HPLC; detection at 258 nm)

Cholesteryl phenylacetate

Cholesteryl phenylacetate ≥98% (HPLC; detection at 258 nm)
Synonym(s):
3β-Hydroxy-5-cholestene 3-phenylacetate, 5-Cholesten-3β-ol 3-phenylacetate
Empirical Formula (Hill Notation):
C35H52O2
CAS Number:
Molecular Weight:
504.79
EC Number:

Products

Product Category
Brand
Biological Source
Formula Weight
Physical Form
Purity
Functional Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing