
Chlorcyclizine hydrochloride

Synonym(s):
1-[(4-Chlorophenyl)phenylmethyl]-4-methylpiperazine monohydrochloride
Empirical Formula (Hill Notation):
C18H21ClN2 · HCl
CAS Number:
Molecular Weight:
337.29
Products
Formula Weight
Manufacturer
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2