Skip to Content
Merck
CHAPS hydrate BioXtra, ≥98% (TLC)

CHAPS hydrate

CHAPS hydrate BioXtra, ≥98% (TLC)
Synonym(s):
3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulfonate hydrate, CHAPS
Empirical Formula (Hill Notation):
C32H58N2O7S · xH2O
CAS Number:
Molecular Weight:
614.88 (anhydrous basis)

Products

Product Category
Brand
Biological Source
Feature
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Tissue
Quality Segment
Markush Class
Markush Group
Technique
Showing 1-3 of 3