
Carbuterol acetate

Empirical Formula (Hill Notation):
C13H21N3O3 · C2H4O2
CAS Number:
Molecular Weight:
327.38
Products
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing